missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium hydroxide, Macron Fine Chemicals™
macron
Supplier: Avantor 325614
Specifications
Ammonium hydroxide | |
H5NO | |
0.91g/cm3 | |
500mL | |
[NH4+].[OH-] | |
35.05 | |
CHEBI:18219 |
1336-21-6 | |
MFCD00066650 | |
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
ammonium hydroxide | |
14923 | |
35.04 |