missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium Hydroxide (Certified ACS Plus), Fisher Chemical™
$205.08 - $12945.85
Chemical Identifiers
CAS | 1336-21-6 |
---|---|
Molecular Formula | H5NO |
Molecular Weight (g/mol) | 35.05 |
MDL Number | MFCD00066650 |
InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
Synonym | ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh |
PubChem CID | 14923 |
ChEBI | CHEBI:18219 |
IUPAC Name | amine hydrate |
SMILES | N.O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
A669-500
|
Fisher Chemical
FLA669500 |
500 mL | Glass Bottle |
Each for $205.08
Case of 6 Each for $1,230.48
|
|
||||
A669S-500
|
Fisher Chemical
FLA669S500 |
500 mL | Safe-Cote™ Glass |
Each for $262.23
Case of 6 Each for $1,573.38
|
|
||||
A669-212
|
Fisher Chemical
FLA669212 |
2.5 L | Glass Bottle |
Each for $542.23
|
|
||||
A669C-212
|
Fisher Chemical
A669C212 |
6 x 2.5 L | Glass Bottle |
Case of 6 Each for $2,257.31
|
|
||||
A669S-212
|
Fisher Chemical
FLA669S212 |
6 x 2.5 L | Safe-Cote™ Glass |
Each for $477.08
Case of 6 Each for $2,862.48
|
|
||||
A669-385LB
|
Fisher Chemical
FLA669385LB |
385 lb. | Poly Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
1336-21-6 | |
35.05 | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
14923 | |
amine hydrate |
H5NO | |
MFCD00066650 | |
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
CHEBI:18219 | |
N.O |
Specifications
12 | |
38°C | |
-57°C | |
H5NO | |
MFCD00066650 | |
500 mL | |
N.O | |
35.05 | |
CHEBI:18219 | |
28.0 to 30.0% (w/w) | |
Pass test | |
Glass Bottle | |
500 hPa at 20°C | |
Ammonium Hydroxide, about 14.8N |
Colorless | |
1336-21-6,7664-41-7,7732-18-5 | |
Assay: 28.0 to 30.0 w/w % | |
NH4OH | |
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
amine hydrate | |
14923 | |
35.05 | |
Certified ACS Plus | |
about 14.8N | |
3ppm max. | |
Liquid |
Safety and Handling
DOTInformation : DOT Class 8: Corrosive