missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium hydroxide, 1.0N Standardized Solution, Thermo Scientific Chemicals
$132.67 - $230.10
Chemical Identifiers
CAS | 1336-21-6 |
---|---|
Molecular Formula | H5NO |
Molecular Weight (g/mol) | 35.05 |
MDL Number | MFCD00066650 |
InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
PubChem CID | 14923 |
ChEBI | CHEBI:18219 |
SMILES | N.O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1336-21-6 | |
35.05 | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
CHEBI:18219 |
H5NO | |
MFCD00066650 | |
14923 | |
N.O |
Specifications
1336-21-6 | |
Colorless | |
MFCD00066650 | |
N.O | |
35.05 | |
CHEBI:18219 | |
Ammonia-like | |
Liquid |
1 L | |
H5NO | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
amine hydrate | |
14923 | |
35.05 | |
23 hPa (17mm Hg) at 20°C | |
Ammonium hydroxide |
Safety and Handling
P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362
H315-H318
EINECSNumber : 215-647-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only