missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium Acetate (Crystalline/Certified ACS), Fisher Chemical™
$320.33 - $2656.50
Chemical Identifiers
CAS | 631-61-8 |
---|---|
Molecular Formula | C2H7NO2 |
Molecular Weight (g/mol) | 77.083 |
MDL Number | MFCD00013066 |
InChI Key | USFZMSVCRYTOJT-UHFFFAOYSA-N |
Synonym | ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 |
PubChem CID | 517165 |
ChEBI | CHEBI:62947 |
IUPAC Name | azanium;acetate |
SMILES | CC(=O)[O-].[NH4+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
A637-500
|
Fisher Chemical
FLA637500 |
500 g | Poly Bottle |
Each for $320.33
Case of 6 Each for $1,921.98
|
|
||||
A637-3
|
Fisher Chemical
FLA6373 |
3 kg | Poly Pail |
Each for $975.00
|
|
||||
A637-10
|
Fisher Chemical
FLA63710 |
10 kg | Poly Pail |
Each for $2,656.50
|
|
||||
Chemical Identifiers
631-61-8 | |
77.083 | |
USFZMSVCRYTOJT-UHFFFAOYSA-N | |
517165 | |
azanium;acetate |
C2H7NO2 | |
MFCD00013066 | |
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 | |
CHEBI:62947 | |
CC(=O)[O-].[NH4+] |
Specifications
114°C | |
White | |
Solid | |
≥97% | |
CH3COONH4 | |
136°C | |
USFZMSVCRYTOJT-UHFFFAOYSA-N | |
azanium;acetate | |
517165 | |
77.08 | |
Certified ACS | |
Poly Bottle | |
Ammonium Acetate |
631-61-8 | |
7.0 | |
500 g | |
C2H7NO2 | |
MFCD00013066 | |
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 | |
CC(=O)[O-].[NH4+] | |
77.083 | |
CHEBI:62947 | |
≥97% | |
Pass Test | |
1.07g/mL |
Safety and Handling
CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:1
Flammability:1
Instability:1