American Scientific Calorimeter
For determining specific heat, heat of fusion and heat of vaporization.
Supplier: American Scientific 323500
Alert:
Intended for children older than 12 years of age.
Description
- Consists of two polished spun aluminum vessels, i.e. an inner calorimeter (2.5D × 3.1 in.. H) and an outer calorimeter (3.9D × 5.5 in. H)
- Includes a plastic ring for seating the inner calorimeter, a stirrer, and plastic lid for outer vessel
- Lid has two holes for the thermometer and the stirrer
- Supplied without the thermometer
Specifications
Calorimeter | |
Determining specific heat, heat of fusion and heat of vaporization | |
2 polished spun aluminum vessels (inner 2.5 D x 3.1 in. H) and outer (3.9 D x 5.5 in. H) calorimeter, plastic ring for seating the inner calorimeter, a stirrer and plastic lid for outer vessel, lid has two holes for the thermometer |