missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ LiCl Precipitation Solution (7.5M) DFS Item

Will not precipitate DNA, protein, or carbohydrates

Manufacturer:  Invitrogen™ LSAM9480

Catalog No. AM9480



Description

Description

Precipitation solution offers major advantages over other RNA precipitation methods in that it does not precipitate DNA, protein, or carbohydrates. It is supplied in one bottle of 100mL at a concentration of 7.5M. It is the method of choice for removing inhibitors of translation or cDNA synthesis from RNA preparations. Note: LiCl precipitation may not remove all DNA contamination from total RNA isolations. If the RNA is to be used for RT-PCR, it should be DNase I treated. The solution is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

DNA and RNA Purification and Analysis, General RNA Purification Reagents and Accessories, RNA Extraction, Total RNA Isolation, Total RNA from Animal Cells and Tissues

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

100mL
Store at 4°C or room temperature.
7.5M
Documents

For Research Use Only. Not for use in diagnostic procedures.