missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Amberlyst™ A-21, ion-exchange resin, Thermo Scientific Chemicals
$426.32 - $426.32
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC202170010
|
Thermo Scientific Chemicals
202170010 |
1 kg | Plastic bottle |
Each for $426.32
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resinChemical Identifiers
9049-93-8 | |
MFCD00145842 |
Specifications
Beads | |
Particle size analysis: uniformity coefficient: <1.80, harmonic mean size: 0.490 to 0.690mm,% <0.300mm: <1.0%,%>1.180mm: <2.0% | |
(free base):> 1.30 eq/L | |
(free base): 52.0 to 62.0% | |
06,302; 07,182; 08,263 | |
1 kg |
9049-93-8 | |
Plastic bottle | |
MFCD00145842 | |
Brown | |
10, 381 | |
Amberlyst™ A-21 |
RUO – Research Use Only