missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Amberlite™ XAD 1180 ion-exchange resin, Thermo Scientific Chemicals
$83.80 - $220.24
Chemical Identifiers
CAS | 9003-69-4 |
---|---|
Molecular Formula | C10H10 |
Molecular Weight (g/mol) | 0.00 |
MDL Number | MFCD00132704 |
InChI Key | MYRTYDVEIRVNKP-UHFFFAOYSA-N |
Synonym | divinylbenzene, 1,2-divinylbenzene, o-divinylbenzene, benzene, diethenyl, benzene, 1,2-diethenyl, divinylbenzen, divinyl benzen, divinyl-benzene, poly divinylbenzene, divinylbenzene dvb |
PubChem CID | 66666 |
SMILES | * |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC299151000
|
Thermo Scientific Chemicals
299151000 |
100 g | Plastic Bottle |
Each for $83.80
|
|
||||
AC299155000
|
Thermo Scientific Chemicals
299155000 |
500 g | Plastic Bottle |
Each for $220.24
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
9003-69-4 | |
0.00 | |
MYRTYDVEIRVNKP-UHFFFAOYSA-N | |
66666 |
C10H10 | |
MFCD00132704 | |
divinylbenzene, 1,2-divinylbenzene, o-divinylbenzene, benzene, diethenyl, benzene, 1,2-diethenyl, divinylbenzen, divinyl benzen, divinyl-benzene, poly divinylbenzene, divinylbenzene dvb | |
* |
Specifications
Beads | |
Particle size analysis: uniformity coefficient: <1.80, harmonic mean size: 0.350 to 0.600mm, <0.250mm: <5.0%,>0.850mm: <10.0% | |
C10H10 | |
moisture holding capacity: 61.0 to 67.0% | |
14, 382 | |
divinylbenzene, 1,2-divinylbenzene, o-divinylbenzene, benzene, diethenyl, benzene, 1,2-diethenyl, divinylbenzen, divinyl benzen, divinyl-benzene, poly divinylbenzene, divinylbenzene dvb | |
* | |
0.00 | |
Amberlite™ XAD 1180 |
9003-69-4 | |
Plastic Bottle | |
MFCD00132704 | |
White to Yellow | |
100 g | |
MYRTYDVEIRVNKP-UHFFFAOYSA-N | |
1,2-diethenylbenzene | |
66666 |
RUO – Research Use Only