Learn More
Aluminum magnesium isopropoxide, 99.9% (metals basis), 10% w/v in isopropanol, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 036591AC
Description
Aluminum magnesium isopropoxide has been used to study the hydrolysis conditions on the properties of magnesium aluminate spinel powders.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Aluminum magnesium isopropoxide | |
Colorless | |
(metals basis) | |
25 mL | |
Moisture sensitive | |
Not miscible with water. | |
99.9% |
69207-83-6 | |
130°C to 142°C (2.0 mmHg) | |
MFCD00070426 | |
UN1219 | |
Magnesium aluminum isopropoxide | |
550.53 | |
Liquid |
Chemical Identifiers
69207-83-6 | |
MFCD00070426 | |
Magnesium aluminum isopropoxide |
Safety and Handling
GHS H Statement
H225-H315-H319-H336
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only