Learn More
Aluminum isopropoxide, 99.99+% (metals basis), Thermo Scientific Chemicals
$173.90 - $528.37
Chemical Identifiers
CAS | 555-31-7 |
---|---|
Molecular Formula | C9H21AlO3 |
Molecular Weight (g/mol) | 204.246 |
MDL Number | MFCD00008870 |
InChI Key | SMZOGRDCAXLAAR-UHFFFAOYSA-N |
Synonym | aluminum isopropoxide, aluminum isopropoxide, aluminum isopropylate, aluminum triisopropoxide, triisopropoxyaluminum, aliso, aluminum triisopropanolate, 2-propanol, aluminum salt, triisopropyloxyaluminum, aluminum triisopropylate |
PubChem CID | 11143 |
IUPAC Name | aluminum;propan-2-olate |
SMILES | CC(C)[O-].CC(C)[O-].CC(C)[O-].[Al+3] |
Description
Aluminum isopropoxide is a useful reagent in the synthesis of alkoxides, chelates and acrylates. It acts as an intermediate of pharmaceutical products like isophytol, testosterone, progesterone and ethisterone. Also used as the material of aluminate coupling agent. It is also used in alcoholysis and ester exchange reactions. In Meerwein-Ponndorf-Verley (MPV) reduction, it is used as a catalyst for the reduction of ketones and aldehydes to their corresponding alcohols. It acts as a precursor to prepare alumina.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
555-31-7 | |
204.246 | |
SMZOGRDCAXLAAR-UHFFFAOYSA-N | |
11143 | |
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Al+3] |
C9H21AlO3 | |
MFCD00008870 | |
aluminum isopropoxide, aluminum isopropoxide, aluminum isopropylate, aluminum triisopropoxide, triisopropoxyaluminum, aliso, aluminum triisopropanolate, 2-propanol, aluminum salt, triisopropyloxyaluminum, aluminum triisopropylate | |
aluminum;propan-2-olate |
Specifications
555-31-7 | |
1.035 g/mL | |
C9H21AlO3 | |
MFCD00008870 | |
UN3181 | |
14,347 | |
Soluble in ethanol,isopropanol,benzene,toluene,chloroform,carbon tetrachloride and petroleum hydrocarbons. | |
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Al+3] | |
204.246 | |
204.25 | |
Powder | |
(metals basis) |
131°C to 135°C | |
135°C (10 mmHg) | |
1.0346 | |
25 g | |
Moisture sensitive | |
aluminum isopropoxide, aluminum isopropoxide, aluminum isopropylate, aluminum triisopropoxide, triisopropoxyaluminum, aliso, aluminum triisopropanolate, 2-propanol, aluminum salt, triisopropyloxyaluminum, aluminum triisopropylate | |
SMZOGRDCAXLAAR-UHFFFAOYSA-N | |
aluminum;propan-2-olate | |
11143 | |
≥99.99% | |
Aluminum isopropoxide |
Safety and Handling
GHS H Statement
H228
Flammable solid.
P210-P240-P241-P261-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
H228-H319-H336
DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
EINECSNumber : 209-090-8
RTECSNumber : BD0975000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only