Learn More
Aluminum isopropoxide, 98+%, Thermo Scientific Chemicals
Reagent for the reduction of carbonyl compounds
$107.33 - $354.48
Chemical Identifiers
CAS | 555-31-7 |
---|---|
Molecular Formula | C9H21AlO3 |
Molecular Weight (g/mol) | 204.246 |
MDL Number | MFCD00008870 |
InChI Key | SMZOGRDCAXLAAR-UHFFFAOYSA-N |
Synonym | aluminum isopropoxide, aluminum isopropoxide, aluminum isopropylate, aluminum triisopropoxide, triisopropoxyaluminum, aliso, aluminum triisopropanolate, 2-propanol, aluminum salt, triisopropyloxyaluminum, aluminum triisopropylate |
PubChem CID | 11143 |
IUPAC Name | aluminum;propan-2-olate |
SMILES | CC(C)[O-].CC(C)[O-].CC(C)[O-].[Al+3] |
Description
Reagent for the reduction of carbonyl compounds. Used in alcoholysis and ester exchange reactions. Also used in synthesis of higher alkoxides, chelates, and acrylates. Aluminum isopropoxide is a useful reagent in the synthesis of alkoxides, chelates and acrylates. It acts as an intermediate of pharmaceutical products like isophytol, testosterone, progesterone and ethisterone. Also used as the material of aluminate coupling agent. It is also used in alcoholysis and ester exchange reactions. In Meerwein-Ponndorf-Verley (MPV) reduction, it is used as a catalyst for the reduction of ketones and aldehydes to their corresponding alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Reagent for the reduction of carbonyl compounds
- Used in alcoholysis and ester exchange reactions
- Also used in synthesis of higher alkoxides, chelates, and acrylates
Chemical Identifiers
555-31-7 | |
204.246 | |
SMZOGRDCAXLAAR-UHFFFAOYSA-N | |
11143 | |
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Al+3] |
C9H21AlO3 | |
MFCD00008870 | |
aluminum isopropoxide, aluminum isopropoxide, aluminum isopropylate, aluminum triisopropoxide, triisopropoxyaluminum, aliso, aluminum triisopropanolate, 2-propanol, aluminum salt, triisopropyloxyaluminum, aluminum triisopropylate | |
aluminum;propan-2-olate |
Specifications
555-31-7 | |
1.035 | |
98+% | |
1.0346 | |
1 kg | |
Moisture sensitive | |
aluminum isopropoxide, aluminum isopropoxide, aluminum isopropylate, aluminum triisopropoxide, triisopropoxyaluminum, aliso, aluminum triisopropanolate, 2-propanol, aluminum salt, triisopropyloxyaluminum, aluminum triisopropylate | |
SMZOGRDCAXLAAR-UHFFFAOYSA-N | |
aluminum;propan-2-olate | |
11143 | |
≥98% | |
Aluminum isopropoxide |
131°C to 135°C | |
135°C (10 mm) | |
C9H21AlO3 | |
MFCD00008870 | |
UN3181 | |
14,347 | |
Soluble in alcohol,benzene,and toluene; Decomposes in water;Soluble in ethanol,isopropanol,benzene,toluene,chloroform,carbon tetrachloride and petroleum hydrocarbons. | |
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Al+3] | |
204.246 | |
204.25 | |
Granules |
Safety and Handling
GHS H Statement
H228
Flammable solid.
P210-P240-P241-P261-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
H228-H319-H336
DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
EINECSNumber : 209-090-8
RTECSNumber : BD0975000
TSCA : Yes
storageNote1 : Moisture Sensitive
Recommended Storage : Ambient temperatures
RUO – Research Use Only