missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum 2,4-pentanedionate, Thermo Scientific Chemicals

$63.34 - $201.08

Chemical Identifiers

CAS 13963-57-0
Molecular Formula C15H21AlO6
Molecular Weight (g/mol) 324.31
MDL Number MFCD00000013
InChI Key KILURZWTCGSYRE-LNTINUHCSA-K
Synonym Aluminum acetylacetonate; Tris(acetylacetonato)aluminum(III)
IUPAC Name aluminium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
SMILES [Al+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1254222
View Documents Promotion Details
Thermo Scientific Chemicals
01254222
100 g
Each for $63.34
 
AA1254236
View Documents Promotion Details
Thermo Scientific Chemicals
01254236
500 g
Each for $201.08
 
Description

Description

Alumunium acetylacetonate may be used to prepare transparent superhydrophobic boehmite and silica films by sublimation, to deposit alumunium oxide films by chemical vapor deposition, to deposit alumunium oxide films by chemical vapor deposition, as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13963-57-0
324.31
KILURZWTCGSYRE-LNTINUHCSA-K
aluminium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
C15H21AlO6
MFCD00000013
Aluminum acetylacetonate; Tris(acetylacetonato)aluminum(III)
[Al+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

13963-57-0
1.27 g/mL
C15H21AlO6
100 g
4157942
It is soluble in organic solvents but insoluble in water.
[Al+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
324.31
Powder
192°C to 194°C
314°C to 316°C
MFCD00000013
UN3467
Aluminum acetylacetonate; Tris(acetylacetonato)aluminum(III)
KILURZWTCGSYRE-LNTINUHCSA-K
aluminium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
324.31
Aluminum 2,4-pentanedionate
Safety and Handling

Safety and Handling

GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H300-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC COMPOUND, SOLID, TOXIC, N.O.S.

EINECSNumber : 237-741-6

RTECSNumber : BD2230000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only