missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
AlphaProTech Critical Cover™ GenPro™ Lab Coats
Soft polypropylene lab coats designed for non-hazardous environments.
$127.65 - $147.34
Specifications
Type | Visitor Lab Coat |
---|---|
Material | Polypropylene |
Color | White |
Closure Type | Snaps |
Cuff Style | Elastic |
Catalog Number | Mfr. No. | Length (English) | Size | Size (English) Chest | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (English) | Size | Size (English) Chest | Price | Quantity | |||
19-130-4241
|
AlphaProTech
LC124214 |
42.75 in. | X-Large | 27.75 in. |
N/A
|
|
|||
19-130-4238
|
AlphaProTech
LC124211 |
36.75 in. | Small | 24.25 in. |
N/A
|
|
|||
19-130-4243
|
AlphaProTech
LC124216 |
44.5 in. | 3X-Large | 30.75 in. |
N/A
|
|
|||
19-130-4239
|
AlphaProTech
LC124212 |
39.5 in. | Medium | 25.25 in. |
N/A
|
|
|||
19-130-4242
|
AlphaProTech
LC124215 |
43.5 in. | 2X-Large | 29.5 in. |
N/A
|
|
|||
19-130-4240
|
AlphaProTech
LC124213 |
40 in. | Large | 27 in. |
N/A
|
|
|||
Description
- Designed for non-hazardous, general purpose environments
- Ideal for personnel as well as facility visitors
- Offer comfort and breathability
- Full length, no-pocket design helps you control contaminants from entering or leaving sensitive areas
Specifications
Visitor Lab Coat | |
White | |
Elastic | |
Non-sterile | |
Tapered | |
Serged |
Polypropylene | |
Snaps | |
Single-use | |
None | |
Yes |