Learn More
Allyl isocyanate, 96%, Thermo Scientific Chemicals
$61.04 - $187.49
Chemical Identifiers
CAS | 1476-23-9 |
---|---|
Molecular Formula | C4H5NO |
Molecular Weight (g/mol) | 83.09 |
MDL Number | MFCD00002044 |
InChI Key | HXBPYFMVGFDZFT-UHFFFAOYSA-N |
PubChem CID | 15123 |
IUPAC Name | 3-isocyanatoprop-1-ene |
SMILES | C=CCN=C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1871003
|
Thermo Scientific Chemicals
L1871003 |
1 g |
N/A
|
|
|||||
AAL1871006
|
Thermo Scientific Chemicals
L1871006 |
5 g |
N/A
|
|
|||||
Description
It finds its application in the synthesis, characterization of biodegradable dextran-allyl isocyanate-ethylamine/polyethylene glycol-diacrylate hydrogels and their in vitro release of albumin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds its application in the synthesis, characterization of biodegradable dextran-allyl isocyanate-ethylamine/polyethylene glycol-diacrylate hydrogels and their in vitro release of albumin.
Solubility
Hydrolyzes in water.
Notes
Moisture sensitive. Store away from alcohols, amines, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away. Protect from electrostatic charges. Store under inert gas.
Chemical Identifiers
1476-23-9 | |
83.09 | |
HXBPYFMVGFDZFT-UHFFFAOYSA-N | |
3-isocyanatoprop-1-ene |
C4H5NO | |
MFCD00002044 | |
15123 | |
C=CCN=C=O |
Specifications
1476-23-9 | |
87°C to 89°C | |
Pungent | |
1.4185 | |
1 g | |
506106 | |
allyl isocyanate, 1-propene, 3-isocyanato, 3-isocyanato-1-propene, propene, 3-isocyanato, allylisocyanate, isocyanic acid, allyl ester, 1-propene,3-isocyanato, prop-2-enisocyanate, 2-propenyl isocyanate, isocyanicacid,allylester | |
HXBPYFMVGFDZFT-UHFFFAOYSA-N | |
3-isocyanatoprop-1-ene | |
15123 | |
98% |
0.94 | |
6°C (43°F) | |
C4H5NO | |
MFCD00002044 | |
UN2478 | |
Moisture sensitive; May form a precipitate | |
Hydrolyzes in water. | |
C=CCN=C=O | |
83.09 | |
83.09 | |
Allyl isocyanate |
Safety and Handling
GHS H Statement
H225-H300-H330-H334-H314-H318
Highly flammable liquid and vapor.
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
H225-H301+H331-H314-H317-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOCYANATES, FLAMMABLE, TOXIC, N.O.S.
EINECSNumber : 000-000-0
RTECSNumber : NQ8175000
TSCA : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only