Sulfite, Quant™ Test Strips, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 023288QA
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
100 strips to measure 10-500ppm SO3â»2 in aqueous solution. | |
100 strips | |
Sulfite, Quant™ Test Strips |
1 kit | |
Ambient temperatures |
RUO – Research Use Only