missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alcian Blue 8GX, certified, ACROS Organics™

$182.68 - $1,206.88

Chemical Identifiers

CAS 33864-99-2
Molecular Formula C56H68Cl4CuN16S4
Molecular Weight (g/mol) 1298.858
MDL Number MFCD00010720
InChI Key CKLBXIYTBHXJEH-UHFFFAOYSA-J
Synonym Ingrain Blue 1
PubChem CID 17749099
SMILES CN(C)C(=[N+](C)C)SCC1=CC2=C(C=C1)C3=NC4=C5C=C(C=CC5=C([N-]4)N=C6C7=C(C=CC(=C7)CSC(=[N+](C)C)N(C)C)C(=N6)N=C8C9=C(C=CC(=C9)CSC(=[N+](C)C)N(C)C)C(=N8)N=C2[N-]3)CSC(=[N+](C)C)N(C)C.[Cl-].[Cl-].[Cl-].[Cl-].[Cu+2]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC400461000 SDS View Documents Acros Organics
AC400461000
100g Glass bottle Each for $1,206.88
AC400460100 SDS View Documents Acros Organics
AC400460100
10g Glass bottle Each for $182.68
AC400460250 SDS View Documents Acros Organics
400460250
25g Glass bottle Each for $393.56
Specifications

Chemical Identifiers

33864-99-2
1298.858
CKLBXIYTBHXJEH-UHFFFAOYSA-J
17749099
C56H68Cl4CuN16S4
MFCD00010720
Ingrain Blue 1
CN(C)C(=[N+](C)C)SCC1=CC2=C(C=C1)C3=NC4=C5C=C(C=CC5=C([N-]4)N=C6C7=C(C=CC(=C7)CSC(=[N+](C)C)N(C)C)C(=N6)N=C8C9=C(C=CC(=C9)CSC(=[N+](C)C)N(C)C)C(=N8)N=C2[N-]3)CSC(=[N+](C)C)N(C)C.[Cl-].[Cl-].[Cl-].[Cl-].[Cu+2]

Specifications

33864-99-2
C56H68Cl4CuN16S4
15, 212
Authentic
CN(C)C(=[N+](C)C)SCC1=CC2=C(C=C1)C3=NC4=C5C=C(C=CC5=C([N-]4)N=C6C7=C(C=CC(=C7)CSC(=[N+](C)C)N(C)C)C(=N6)N=C8C9=C(C=CC(=C9)CSC(=[N+](C)C)N(C)C)C(=N8)N=C2[N-]3)CSC(=[N+](C)C)N(C)C.[Cl-].[Cl-].[Cl-].[Cl-].[Cu+2]
17749099
Crystalline Powder
Solubility in water: moderately soluble. Other solubilities: soluble in ethanol, cellosolve, ethylene glycol, practically insoluble in xylene
Purple
Alcian Blue 8GX
50% min.
MFCD00010720
Ingrain Blue 1
CKLBXIYTBHXJEH-UHFFFAOYSA-J
1298.858
1298.88
Glass bottle
148.0°C
100g