missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Albumin, Human, Fraction V Protease, HRP, and AP Free, 10g, MP Biomedicals

Manufacturer:  MP Biochemicals LLC 08823002

Catalog No. ICN823002



Specifications

Specifications

1mg/mL
Powder
6.8
2° to 8°C
Immunoassays, Mammalian Cell Culture
Human Albumin Fraction V
10g
Documents