AirClean™ Systems Activated Bonded Carbon Chemisorptive Filters
For the removal of fumes and vapors
$1080.95 - $1270.58
Catalog Number | Mfr. No. | Filter Type | For Use With (Application) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Filter Type | For Use With (Application) | Price | Quantity | ||||
36-101-8827
|
AirClean™ Systems
ACF300 |
ACF300 | Ammonia or Amines |
Each for $1,194.72
|
|
||||
36-101-8825
|
AirClean™ Systems
ACF100 |
ACF100 | Organic Vapor, Solvents, and Odors |
Each for $1,080.95
|
|
||||
36-101-8828
|
AirClean™ Systems
ACF400 |
ACF400 | Inorganic Acids |
Each for $1,194.72
|
|
||||
361018829
|
AirClean™ Systems
ACFFOR |
ACFFOR | Formaldehyde |
Each for $1,210.98
|
|
||||
361018843
|
AirClean™ Systems
ACF100H |
ACF100H | Organic Solvent Vapors |
Each for $1,270.58
|
|
||||
Description
- Coconut shell activated carbon
- Chemically impregnated carbon for increased load capacity
- Large surface area and highly developed internal structure allows adsorption up to 50%
- Bonded design eliminates carbon dusting and prevents carbon shift that can lead to “dead spots”
Specifications
Activated Carbon |