missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals Agarose Gel Loading Dye (6X, Ficoll-Based)
$213.05 - $213.05
Specifications
Chemical Name or Material | Agarose gel loading dye |
---|---|
Physical Form | Viscous Liquid |
Name Note | Ficoll based |
Concentration | 6X |
MDL Number | MFCD00675058 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ62800AC
|
Thermo Scientific Chemicals
J62800AC |
25 mL |
Each for $213.05
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Agarose gel loading dye | |
Ficoll based | |
MFCD00675058 |
Viscous Liquid | |
6X |
RUO – Research Use Only