missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Adapters for Round Single Blood Bag Buckets for Thermo Scientific™ HAEMAFlex™ 6/8 Rotors
Select adapters for 450-500mL blood bags in round single blood bag buckets for Thermo Scientific™ HAEMAFlex™ 6/8 rotors.
$499.72 - $546.57
Specifications
Type | Blood Bag Adapter |
---|---|
For Use With (Equipment) | Single blood bag bucket with filter pack |
Catalog Number | Mfr. No. | Description | Capacity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Capacity | Price | Quantity | ||||
75-003-858
|
Thermo Scientific™
75003858 |
450mL Single or Double Blood Bag Adapter (set of 2) | 6/8 x 450 mL |
Pack of 2 Each for $499.72
|
|
||||
75-003-857
|
Thermo Scientific™
75003857 |
450-500mL Triple or Quad Blood Bag Adapter (set of 2) | 6/8 x 500 mL |
Pack of 2 Each for $546.57
|
|
||||
Description
Thermo Scientific™ blood banking centrifuges offer the flexibility to select higher capacity rotors or match existing workflows, with a choice of Thermo Scientific™ HAEMAFlex™ rotors and bucket systems.
Address your lab’s regular- to medium-throughput blood processing needs with the HAEMAFlex 6 swinging bucket rotor or, utilizing the same bucket system, upgrade to the HAEMAFlex 8 swinging bucket rotor.Select adapters for 450–500mL Triple or Quad blood bags or 450mL Single or Double blood bags.
Specifications
Blood Bag Adapter | |
Single blood bag bucket with filter pack |