Learn More
Acetyl chloride, 99+%, Thermo Scientific Chemicals
Acetyl chloride, 99+%, C2H3ClO, CAS Number-75-36-5, acetic acid chloride, acetic acid, chloride, acetic chloride, acetylchloride, ccris 4568, ch3cocl, ethanoyl chloride, hsdb 662, rcra waste number u006, unii-qd15rno45k, 250mL, 99% min. (GC), 02, 173, 51.0 deg.C, 100.0, 99.0, CHEBI:37580
$79.70 - $229.72
Chemical Identifiers
CAS | 75-36-5 |
---|---|
Molecular Formula | C2H3ClO |
Molecular Weight (g/mol) | 78.50 |
MDL Number | MFCD00000719 |
InChI Key | WETWJCDKMRHUPV-UHFFFAOYSA-N |
Synonym | ethanoyl chloride, acetic chloride, acetylchloride, acetic acid chloride, ch3cocl, rcra waste number u006, acetic acid, chloride, unii-qd15rno45k, ccris 4568, hsdb 662 |
PubChem CID | 6367 |
ChEBI | CHEBI:37580 |
IUPAC Name | acetyl chloride |
SMILES | CC(Cl)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC219470250
|
Thermo Scientific Chemicals
219470250 |
25 mL | Glass bottle |
Each for $79.70
|
|
||||
AC219472500
|
Thermo Scientific Chemicals
219472500 |
250 mL | Glass bottle |
Each for $95.65
|
|
||||
AC219470010
|
Thermo Scientific Chemicals
219470010 |
1 L | Glass bottle |
Each for $229.72
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-36-5 | |
78.50 | |
WETWJCDKMRHUPV-UHFFFAOYSA-N | |
6367 | |
acetyl chloride |
C2H3ClO | |
MFCD00000719 | |
ethanoyl chloride, acetic chloride, acetylchloride, acetic acid chloride, ch3cocl, rcra waste number u006, acetic acid, chloride, unii-qd15rno45k, ccris 4568, hsdb 662 | |
CHEBI:37580 | |
CC(Cl)=O |
Specifications
75-36-5 | |
100.0 | |
1.1000g/mL | |
4°C | |
99% min. (GC) | |
C2H3ClO | |
CH3COCl | |
25 mL | |
01,11; 04,5; 05,4; 07,3; 11,4 | |
15, 80 | |
Solubility in water: reacts. Other solubilities: miscible with acetone, benzenz, toluene,, chloroform, ether, petroleum ether, carbon dosulfi, de and glacial acetic acid, soluble in ccl4, reacts with alcohol | |
CC(Cl)=O | |
78.50 | |
CHEBI:37580 | |
99+% | |
Acetyl chloride, p.a. |
99.0 | |
-112.0°C | |
51.0°C | |
Authentic | |
Glass bottle | |
1.3876 to 1.3896 | |
MFCD00000719 | |
02, 173 | |
1.1 | |
ethanoyl chloride, acetic chloride, acetylchloride, acetic acid chloride, ch3cocl, rcra waste number u006, acetic acid, chloride, unii-qd15rno45k, ccris 4568, hsdb 662 | |
WETWJCDKMRHUPV-UHFFFAOYSA-N | |
acetyl chloride | |
6367 | |
78.5 | |
Liquid |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Reacts violently with water.
Corrosive to the respiratory tract.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Danger
EINECSNumber : 200-865-6
RTECSNumber : AO6390000
TSCA : TSCA
RUO – Research Use Only