Learn More
Acetyl chloride, 99+%, Thermo Scientific Chemicals
Acetyl chloride, 99+%, C2H3ClO, CAS Number-75-36-5, acetic acid chloride, acetic acid, chloride, acetic chloride, acetylchloride, ccris 4568, ch3cocl, ethanoyl chloride, hsdb 662, rcra waste number u006, unii-qd15rno45k, 250mL, 99% min. (GC), 02, 173, 51.0 deg.C, 100.0, 99.0, CHEBI:37580
Supplier: Thermo Scientific Chemicals 219472500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Acetyl chloride | |
75-36-5 | |
100.0 | |
1.1000g/mL | |
4°C | |
99% min. (GC) | |
C2H3ClO | |
CH3COCl | |
250 mL | |
01,11; 04,5; 05,4; 07,3; 11,4 | |
15, 80 | |
Solubility in water: reacts. Other solubilities: miscible with acetone, benzenz, toluene,, chloroform, ether, petroleum ether, carbon dosulfi, de and glacial acetic acid, soluble in ccl4, reacts with alcohol | |
CC(Cl)=O | |
78.50 | |
CHEBI:37580 | |
99+% |
p.a. | |
99.0 | |
-112.0°C | |
51.0°C | |
Authentic | |
Glass bottle | |
1.3876 to 1.3896 | |
MFCD00000719 | |
02, 173 | |
1.1 | |
ethanoyl chloride, acetic chloride, acetylchloride, acetic acid chloride, ch3cocl, rcra waste number u006, acetic acid, chloride, unii-qd15rno45k, ccris 4568, hsdb 662 | |
WETWJCDKMRHUPV-UHFFFAOYSA-N | |
acetyl chloride | |
6367 | |
78.5 | |
Liquid |
Chemical Identifiers
75-36-5 | |
78.50 | |
WETWJCDKMRHUPV-UHFFFAOYSA-N | |
6367 | |
acetyl chloride |
C2H3ClO | |
MFCD00000719 | |
ethanoyl chloride, acetic chloride, acetylchloride, acetic acid chloride, ch3cocl, rcra waste number u006, acetic acid, chloride, unii-qd15rno45k, ccris 4568, hsdb 662 | |
CHEBI:37580 | |
CC(Cl)=O |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Reacts violently with water.
Corrosive to the respiratory tract.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Danger
EINECSNumber : 200-865-6
RTECSNumber : AO6390000
TSCA : TSCA
RUO – Research Use Only