missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetonitrile E CHROMASOLV™, for HPLC, for UV, ≥99.9% (GC), Honeywell Riedel-de Haën™
CHROMASOLV™, for HPLC, for UV,≥99.9% (GC)
$294.55 - $4002.50
Chemical Identifiers
CAS | 75-05-8 |
---|---|
Molecular Formula | C2H3N |
MDL Number | MFCD00001878 |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
PubChem CID | 6342 |
ChEBI | CHEBI:38472 |
IUPAC Name | acetonitrile |
SMILES | CC#N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
6000236
|
Honeywell Chemicals
348881L |
1 L |
Each for $294.55
|
|
|||||
6000237
|
Honeywell-Chromasolv
3488825L |
2.5 L |
Each for $667.45
|
|
|||||
6000238
|
Honeywell-Chromasolv
348884X25L |
4 x 2.5 L |
Case of 4 Each for $2,321.53
|
|
|||||
60048230
|
Honeywell Chemicals
348884X4L |
4 x 4 L |
Case of 4 Each for $4,002.50
|
|
|||||
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Chemical Identifiers
75-05-8 | |
MFCD00001878 | |
6342 | |
acetonitrile |
C2H3N | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
CHEBI:38472 | |
CC#N |
Specifications
-48°C | |
0.786g/mL at 25°C(lit.) | |
1 L | |
C2H3N | |
MFCD00001878 | |
741857 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
41.05g/mol | |
CHROMASOLV™ |
75-05-8 | |
81°C to 82°C | |
n20/D 1.344 | |
CH3CN | |
72.8mmHg (20°C) | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CC#N | |
41.053 | |
CHEBI:38472 | |
≥99.9% (GC) | |
Acetonitrile, for HPLC, for UV |
Safety and Handling
P210-P280-P305 + P351 + P338
H225-H302 + H312 + H332-H319
EINECSNumber : 200-835-2
RTECSNumber : AL7700000