missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetonitrile, Anhydrous, Biosynthesis grade, 99.9% min., OmniSolv, MilliporeSigma™
Supplier: Technidata AX01511
Specifications
Acetonitrile | |
75-05-8 | |
4 L | |
99.9% min. | |
CH3CN | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
41.5 |
OmniSolv™ | |
Colorless | |
Liquid | |
C2H3N | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CC#N | |
41.053 | |
CHEBI:38472 | |
Biosynthesis |
Safety and Handling
Recommended Storage : Room Temperature