missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetonitrile, 99+%, for spectroscopy, Thermo Scientific Chemicals

$108.99 - $465.83

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC167652500
SDS View Documents Promotion Details
Thermo Scientific Chemicals
167652500
250 mL Glass Bottle
Each for $108.99
Case of 6 Each for $653.94
 
AC167650010
SDS View Documents Promotion Details
Thermo Scientific Chemicals
167650010
1 L Glass Bottle
Each for $241.89
Case of 4 Each for $967.56
 
AC167650025
SDS View Documents Promotion Details
Thermo Scientific Chemicals
167650025
2.5 L Glass Bottle
Each for $465.83
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
Specifications

Chemical Identifiers

75-05-8
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
acetonitrile
C2H3N
MFCD00001878
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
CHEBI:38472
CC#N

Specifications

-46.0°C
0.01 max. at 250nm, 0.02 max. at 235nm, 0.1 max. at 220nm, 0.3 max. at 210nm, 1 max. at 193nm
100.0
0.7810g/mL
250 mL
Glass Bottle
1.3430 to 1.3450
99+%
C2H3N
MFCD00001878
0.36 mPa.s (20°C)
02, 183
15, 69
Solubility in water: miscible. Other solubilities: soluble in alcohol, ethyl acetate, acetone, ether
CC#N
41.053
CHEBI:38472
≥99%
Acetonitrile
75-05-8
99.0
>500°C
81.0°C to 82.0°C
2°C
Liquid
0.0003% max.
0.781
CH3CN
Vapor Pressure: 97mbar at 20°C
0.03% max.
02,13; 12,450; 13,156; 15,1
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
6342
41.05
Spectroscopy
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only