missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetonitrile, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™

$78.08 - $322.22

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitril, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC364310025 SDS View Documents Acros Organics
AC364310025
2.5L Glass bottle Each for $322.22
AC364310010 SDS View Documents Acros Organics
364310010
1L AcroSeal™ Glass Bottle Each for $146.80
AC364311000 SDS View Documents Acros Organics
364311000
100mL AcroSeal™ Glass Bottle Each for $78.08
Specifications

Chemical Identifiers

75-05-8
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
acetonitrile
C2H3N
MFCD00001878
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitril, cyanure de methyl, methylkyanid, mecn
CHEBI:38472
CC#N

Specifications

75-05-8
100.0
Vapor Pressure: 97mbar at 20°C
C2H3N
MFCD00001878
02,13; 12,450; 13,156; 15,1
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitril, cyanure de methyl, methylkyanid, mecn
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
6342
41.05
99.9%
0.7810g/mL
1.3430 to 1.3450
0.781
2°C
>500°C
Undesignated
2.5L
99.85
0.01% max.
99.9%
CH3CN
02, 183
15, 69
Solubility in water: miscible. Other solubilities: soluble in alcohol, ethyl acetate, acetone, ether
CC#N
41.053
CHEBI:38472
Liquid
Extra Dry over Molecular Sieve
Glass bottle
Solubility in water: miscible. Other solubilities: soluble in alcohol, ethyl acetate, acetone, ether
0.005% max. (Coulometric)
0.36 mPa.s (20°C)
81.0°C to 82.0°C
-46.0°C
Acetonitrile, water <50 ppm, extra dry over mol. sieve
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger