missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetonitrile, 99.9%, Extra Dry, AcroSeal™, ACROS Organics™

Manufacturer:  Acros Organics 610220010

 View more versions of this product

Catalog No. AC610220010



Specifications

Specifications

Acetonitrile
Vapor Pressure: 97mbar at 20°C
C2H3N
MFCD00001878
02, 183
15, 69
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
6342
41.04
Extra Dry
Pass Test
AcroSeal™ Glass Bottle in Tin Can
50ppm max.
>500°C
-46°C
75-05-8
99.9%
CH3CN
1648; Colorless
02,13; 12,450; 13,156; 15,1
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitril, cyanure de methyl, methylkyanid, mecn
CC#N
41.053
CHEBI:38472
≥99.9%
0.7810g/mL
Conforms to reference
1ppm max.
2°C (35.8°F)
81°C to 82°C
1L

Chemical Identifiers

75-05-8
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
acetonitrile
C2H3N
MFCD00001878
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitril, cyanure de methyl, methylkyanid, mecn
CHEBI:38472
CC#N
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 200-835-2

RTECSNumber : AL7700000

TSCA : TSCA

SDS Documents