missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetonitrile 50% and Water 50% (wt/wt), LabReady™ Solvent Blend, Honeywell Burdick & Jackson
Supplier: Honeywell-Burdick and Jackson 4574
Specifications
Acetonitrile 50% and Water 50% (wt/wt) | |
75-05-8,7732-18-5 | |
56.1000%,50.0000% | |
82°C | |
3°C | |
C2H3N | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CC#N | |
41.053 | |
CHEBI:38472 |
LabReady™ Solvent Blend | |
50.0000%,43.9000% | |
0.7857 g/cm3 | |
4 x 4 L | |
Glass Bottle | |
UN1648 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
Solvent Blend |
Chemical Identifiers
75-05-8 | |
41.053 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CHEBI:38472 | |
CC#N |
C2H3N | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
6342 | |
acetonitrile |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile solution