missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetone, Technical, Fisher Chemical

$269.52 - $1,792.41

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A1320 SDS View Documents Thermo Scientific
A1320
20L Each for $269.52
A13200 SDS View Documents Thermo Scientific
A13200
200L Each for $1,792.41
Specifications

Chemical Identifiers

67-64-1
58.08
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CHEBI:15347
CC(=O)C
C3H6O
CSCPPACGZOOCGX-UHFFFAOYSA-N
180
propan-2-one

Specifications

67-64-1
C3H6O
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CC(=O)C
58.08
CHEBI:15347
Liquid
Technical
Pass Test
Steel pail
Soluble
2.0
0.32mPa.s at 20°C
Undesignated
7
Acetone
Clear, colorless liquid
1090
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
180
58.08
≤98%
0.790
Sweet
1.358 to 1.359
247mbar at 20°C
-20°C (-4°F); Closed cup
56°C
-95°C
20L
Safety and Handling

Safety and Handling

Recommended Storage :

  • Flammables area
  • Keep containers tightly closed in a dry, cool and well-ventilated place
  • Keep away from heat and sources of ignition
  • Keep container tightly closed in a dry and well-ventilated place