missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetone, Histology grade, 99% min, EMD Millipore

$123.60 - $4,704.40

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(=O)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
MAX012520 View Documents Millipore Sigma
AX012520
200L Metal Drum Each for $4,704.40
MAX01256 View Documents Millipore Sigma
AX01256
500mL Glass Bottle Each for $123.60
MAX01254 View Documents Millipore Sigma
AX01254
4L Plastic Bottle Each for $210.34
Specifications

Chemical Identifiers

67-64-1
58.08
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CHEBI:15347
CC(=O)C
C3H6O
CSCPPACGZOOCGX-UHFFFAOYSA-N
180
propan-2-one

Specifications

67-64-1
C3H6O
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
180
58.08
≥99% (GC)
Conforms to standard
To pass test
Colorless
Acetone, For analysis Histology
99% min.
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CC(=O)C
58.08
CHEBI:15347
Liquid
To pass test
Metal Drum
0.001% max
200L
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature