Learn More
Acetone, Environmental Grade, 99.5+%, Thermo Scientific Chemicals
Suitable For Pesticide Residue Analysis
$79.70 - $550.48
Chemical Identifiers
CAS | 67-64-1 |
---|---|
Molecular Formula | C3H6O |
Molecular Weight (g/mol) | 58.08 |
MDL Number | MFCD00008765 |
InChI Key | CSCPPACGZOOCGX-UHFFFAOYSA-N |
Synonym | acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone |
PubChem CID | 180 |
ChEBI | CHEBI:15347 |
IUPAC Name | propan-2-one |
SMILES | CC(C)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA43053K2
|
Thermo Scientific Chemicals
043053K2 |
1 L |
Each for $79.70
|
|
|||||
AA43053K7
|
Thermo Scientific Chemicals
043053K7 |
4 L |
Each for $175.51
|
|
|||||
AA43053M6
|
Thermo Scientific Chemicals
043053M6 |
4 x 4 L |
Each for $550.48
|
|
|||||
Description
Suitable for pesticide residue analysisAcetone is a polar, aprotic solvent used in the synthesis and isolation of both organic and inorganic compounds and complexes. It plays a vital role in Jones oxidation. It is utilized for rinsing laboratory glassware due to its low cost and volatility. It is used in the manufacture of various chemicals such as methyl isobutyl ketone, mesityl oxide, diacetone alcohol, acetic acid, iodoform, bromoform, rayon and photographic films, paint and varnish removers. It is also useful in the synthesis of t-butanesulfinamide, terpenes, thiol-stabilized gold colloids, calixphyrin macrocycles and trispyrazolylborate platinum compounds. It plays an important role in protein crystallization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
67-64-1 | |
58.08 | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
180 | |
propan-2-one |
C3H6O | |
MFCD00008765 | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CHEBI:15347 | |
CC(C)=O |
Specifications
°C to 94°C | |
Colorless | |
56°C | |
-17°C (1°F) | |
Liquid | |
>99.5% | |
MFCD00008765 | |
1090; Colorless | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
propan-2-one | |
180 | |
58.08 | |
Acetone |
67-64-1 | |
0.791 | |
1 L | |
465°C (869°F) | |
1.359 | |
C3H6O | |
185mm Hg at 20°C | |
635680 | |
Miscible with water,benzene,alcohol,dimethylformamide,ether,chloroform and most oils. | |
CC(C)=O | |
58.08 | |
CHEBI:15347 | |
Environmental |
Safety and Handling
Keep container in a well ventilated place. Keep away from sources of ignition - No Smoking. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. If swallowed seek medical advice immediately and show this container or label.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c
H225-H319-H336-H373
DOTInformation : DOT Class: 3, Packaging Group: II
EINECSNumber : 200-662-2
RTECSNumber : AL3150000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only