missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetone, ACS, 99.5+%, Alfa Aesar

$22.99 - $149.45

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(=O)C
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA30698AB SDS View Documents Alfa Aesar
30698AB
10mL Each for $22.99
AA30698AP SDS View Documents Alfa Aesar
30698AP
500mL Each for $34.07
AA30698K2 SDS View Documents Alfa Aesar
30698K2
1L Each for $48.27
AA30698K7 SDS View Documents Alfa Aesar
30698K7
4L Each for $115.51
AA30698M1 SDS missing translation for 'viewDocuments' Alfa Aesar
30698M1
4 x 1L Each for $149.45
Specifications

Chemical Identifiers

67-64-1
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
180
propan-2-one
C3H6O
MFCD00008765
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CHEBI:15347
CC(=O)C

Specifications

67-64-1
>99.5%
MFCD00008765
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CC(=O)C
58.08
CHEBI:15347
Liquid
Characteristic
Colorless
Acetone
Colorless liquid
C3H6O
1090; Colorless
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
180
58.08
ACS
247 hPa (185mm Hg) at 20°C
10mL
Safety and Handling

Safety and Handling

H225-H319-H336: Highly flammable liquid and vapour. Causes serious eye irritation. May cause drowsiness or dizziness.

DOTInformation : DOT Class: 3; Packing Group: II

EINECSNumber : 200-662-2