missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetic acid, 1.0N Standardized Solution, Thermo Scientific Chemicals
$97.66 - $97.66
Chemical Identifiers
CAS | 64-19-7 |
---|---|
Molecular Formula | C2H4O2 |
Molecular Weight (g/mol) | 60.05 |
MDL Number | MFCD00036152 |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
PubChem CID | 176 |
ChEBI | CHEBI:15366 |
IUPAC Name | acetic acid |
SMILES | CC(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA35566K2
|
Thermo Scientific Chemicals
035566K2 |
1 L |
Each for $97.66
|
|
|||||
Description
Acetic acid, 1.0N standardized solution is used for volumetric analysis in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
64-19-7 | |
60.05 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
CHEBI:15366 | |
CC(O)=O |
C2H4O2 | |
MFCD00036152 | |
176 | |
acetic acid |
Specifications
64-19-7 | |
C2H4O2 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
acetic acid | |
176 | |
60.05 | |
Liquid |
1 L | |
MFCD00036152 | |
CC(O)=O | |
60.05 | |
CHEBI:15366 | |
Miscible with water | |
Acetic acid |
Safety and Handling
EINECSNumber : 200-580-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only