Thermo Scientific™ Accessories for Thermo Scientific™ Cimarec+ and SuperNuova+ Series Hotplates
Add functionality to your Thermo Scientific™ Cimarec+ and SuperNuova+ Series Hotplates with these accessories.
$98.22 - $155.08
Products 3
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
CIC0001447 View Documents Thermo Scientific™
CIC0001447
PT100 External probe, 0-400°C, Stainless steel, for all sizes of Cimarec+ and SuperNuova+ Hotplates and Hotplate Stirrers Each for $98.22
CIC0001448 View Documents Thermo Scientific™
CIC0001448
PT1000 External probe, 0-400°C, Stainless steel, for all sizes of Cimarec+ and SuperNuova+ Hotplates and Hotplate Stirrers Each for $105.07
CIC0001449 View Documents Thermo Scientific™
CIC0001449
Hastelloy PT1000 External probe, 0-400°C, Hastelloy, for all sizes of Cimarec+ and SuperNuova+ Hotplates and Hotplate Stirrers Each for $155.08