Abnova™ Nitrate/Nitrite Colorimetric Assay Kit
Provides an accurate and convenient method for measurement of total nitrate/nitrite concentration in a simple two-step process.
Supplier: Abnova™ KA1342
Description
- First step is the conversion of nitrate to nitrite utilizing nitrate reductase
- Second step is the addition of the Griess Reagents which convert nitrite into a deep purple azo compound
- Photometric measurement of the absorbance due to this azo chromophore accurately determines NO2- concentration
Specifications
Store the kit at -20°C temporarily. For best results, store the components as described in the protocol. | |
Colorimetric | |
1 Vial Nitrate/Nitrite Assay Buffer, 2 vials Nitrate Reductase Enzyme preparation, 2 vials Nitrate Reductase Co-factor preparation, 1 vial Nitrate standard, 1 vial Nitrite standard, 2 vials Griess reagent R1, 2 vials Griess reagent R2, 3 plates 96-well solid plate (colorimetric assay), 3 covers 96-well cover sheet | |
Detection Assay Kit |
Nitrate/Nitrite Colorimetric Assay Kit is a measurement for detecting total nitratr/nitrite concentration. | |
Functional Study | |
1 Kit |