missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Abnova™ Cortisol ELISA Kit
For the quantitative measurement of free Cortisol in Urine
Supplier: Abnova™ KA3382
Specifications
0 to 3200pg/mL | |
Cortisol ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative detection of cortisol in dried fecal extracts, saliva, urine, serum, plasma and tissue culture media samples. | |
Colorimetric | |
96 Assays | |
Dried Fecal Extract, Plasma, Saliva, Serum, Tissue Culture Medium Sample, Urine | |
Store the kit at 4°C | |
Cortisol ELISA Kit |
HRP | |
45.4 pg/mL | |
96 wells Clear coated plate, Cortisol Standard (125uL), Cortisol Antibody (3mL), Cortisol Conjugate (3mL), Assay Buffer (28mL), Dissociation Reagent (1mL), Wash Buffer Concentrate (30mL), TMB Substrate (11mL), Stop Solution (5mL), Plate Sealer | |
For research use only (RUO) | |
50 μL | |
Quantification | |
96 |