missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Abnova™ 8-hydroxy-2'-deoxyguanosine ELISA Kit
Use to measure quantitatively 8-hydroxy-2'-deoxyguanosine. Sandwich enzyme immunoassay.
Supplier: Abnova™ KA0444
Includes:
- 8-OHdG Microtiter Plate: Precoated With 8-OHdG (8 x12 wells, Split Type), 1 Plate
- Primary Antibody: Monoclonal Antibody Specific For 8-OHdG
- Primary Antibody Dilution Buffer: Phosphate Buffered Saline
- Secondary Antibody: HRP-Conjugated Antibody
- Secondary Antibody Dilution Buffer: Phosphate Buffered Saline
- Chromogen: 3,3',5,5'-Tetramethylbenzidine
- Chromogen Dilution Buffer: Hydrogen Peroxide/Citrate-Phosphate Buffered Saline
- Washing Buffer (5x): Concentrated Phosphate Buffered Saline, 2 at 26mL
- Stop Solution: 1M Phosphoric Acid
- Standards 1-6
- Plate Seal: Adhesive sheet for covering plate
Description
This Kit is a competitive enzyme-linked immunosorbent assay (ELISA) for quantitative measurement of 8-hydroxy-2â-deoxyguanosine (8-OHdG) in tissue, serum, plasma and urine resulting from oxidative damage to DNASpecifications
0.5 to 200ng/mL | |
8-hydroxy-2'-deoxyguanosine ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of 8-hydroxy-2'-deoxyguanosine. | |
1 Kit | |
Plasma, Serum, Tissue, Urine | |
Store the kit at 4°C | |
8-hydroxy-2'-deoxyguanosine ELISA Kit |
HRP | |
Colorimetric | |
For research use only (RUO) | |
50 μL | |
Quantification | |
96 |