Learn More
6-Chloro-7-deazapurine, 98%, Thermo Scientific Chemicals
$373.36 - $1447.15
Chemical Identifiers
CAS | 3680-69-1 |
---|---|
Molecular Formula | C6H4ClN3 |
Molecular Weight (g/mol) | 153.569 |
MDL Number | MFCD01686865 |
InChI Key | BPTCCCTWWAUJRK-UHFFFAOYSA-N |
Synonym | 4-chloro-7h-pyrrolo 2,3-d pyrimidine, 4-chloropyrrolo 2,3-d pyrimidine, 6-chloro-7-deazapurine, 4-chloro-1h-pyrrolo 2,3-d pyrimidine, 7h-pyrrolo 2,3-d pyrimidine, 4-chloro, 1h-pyrrolo 2,3-d pyrimidine, 4-chloro, 6-chloro-7-deazapurine 4-chloro-1h-pyrrolo 2,3-d pyrimidine, 4-chloro-1h-pyrrol 2,3-d pyrimidine |
PubChem CID | 5356682 |
IUPAC Name | 4-chloro-7H-pyrrolo[2,3-d]pyrimidine |
SMILES | C1=CNC2=C1C(=NC=N2)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2571303
|
Thermo Scientific Chemicals
B2571303 |
1 g |
Each for $373.36
|
|
|||||
AAB2571306
|
Thermo Scientific Chemicals
B2571306 |
5 g |
Each for $1,447.15
|
|
|||||
Description
6-Chloro-7-deazapurine is used for making tofacitinib.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-Chloro-7-deazapurine is used for making tofacitinib.
Solubility
Soluble in DMSO, ethyl acetate and methanol.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
3680-69-1 | |
153.569 | |
BPTCCCTWWAUJRK-UHFFFAOYSA-N | |
5356682 | |
C1=CNC2=C1C(=NC=N2)Cl |
C6H4ClN3 | |
MFCD01686865 | |
4-chloro-7h-pyrrolo 2,3-d pyrimidine, 4-chloropyrrolo 2,3-d pyrimidine, 6-chloro-7-deazapurine, 4-chloro-1h-pyrrolo 2,3-d pyrimidine, 7h-pyrrolo 2,3-d pyrimidine, 4-chloro, 1h-pyrrolo 2,3-d pyrimidine, 4-chloro, 6-chloro-7-deazapurine 4-chloro-1h-pyrrolo 2,3-d pyrimidine, 4-chloro-1h-pyrrol 2,3-d pyrimidine | |
4-chloro-7H-pyrrolo[2,3-d]pyrimidine |
Specifications
3680-69-1 | |
C6H4ClN3 | |
1 g | |
Soluble in DMSO,ethyl acetate and methanol. | |
C1=CNC2=C1C(=NC=N2)Cl | |
153.569 | |
153.52 | |
6-Chloro-7-deazapurine |
190°C to 192°C | |
MFCD01686865 | |
4-chloro-7h-pyrrolo 2,3-d pyrimidine, 4-chloropyrrolo 2,3-d pyrimidine, 6-chloro-7-deazapurine, 4-chloro-1h-pyrrolo 2,3-d pyrimidine, 7h-pyrrolo 2,3-d pyrimidine, 4-chloro, 1h-pyrrolo 2,3-d pyrimidine, 4-chloro, 6-chloro-7-deazapurine 4-chloro-1h-pyrrolo 2,3-d pyrimidine, 4-chloro-1h-pyrrol 2,3-d pyrimidine | |
BPTCCCTWWAUJRK-UHFFFAOYSA-N | |
4-chloro-7H-pyrrolo[2,3-d]pyrimidine | |
5356682 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only