Learn More
Thermo Scientific™ 5000 Series 0.31μm to 0.93μm Polymer Particle Suspensions
Meet the need for particulate materials with a variety of particle sizes and properties with Thermo Scientific™ 5000 Series Polymer Particle Suspensions
$1189.53 - $30084.97
Specifications
Additive | Contains trace amount of surfactant to inhibit agglomeration and promote stability |
---|---|
Concentration | 10% |
Content And Storage | Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer. |
Description | Particles useful for such applications as filter evaluation, checking and testing, light scattering research, fluid mechanics research, aerosol particle generation, dispersion studies, and many other research and development projects. They are not intended for use in instrument calibration. |
Color | White or Colorless |
Catalog Number | Mfr. No. | Diameter (Metric) | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Quantity | Price | Quantity | ||||
09980284
|
Thermo Scientific™
5031A |
0.31 μm | 15 mL |
Each for $1,189.53
|
|
||||
09980285
|
Thermo Scientific™
5031B |
0.31 μm | 100 mL |
Each for $4,644.38
|
|
||||
09980286
|
Thermo Scientific™
5031C |
0.31 μm | 1000 mL |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
09980287
|
Thermo Scientific™
5033A |
0.33 μm | 15 mL |
Each for $1,189.53
|
|
||||
09980288
|
Thermo Scientific™
5033B |
0.33 μm | 100 mL |
Each for $4,644.38
|
|
||||
09980289
|
Thermo Scientific™
5033C |
0.33 μm | 1000 mL |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
09980290
|
Thermo Scientific™
5036A |
0.36 μm | 15 mL |
Each for $1,189.53
|
|
||||
09980291
|
Thermo Scientific™
5036B |
0.36 μm | 100 mL |
Each for $4,644.38
|
|
||||
09980292
|
Thermo Scientific™
5036C |
0.36 μm | 1000 mL |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
09980293
|
Thermo Scientific™
5043A |
0.43 μm | 15 mL |
Each for $1,189.53
|
|
||||
09980294
|
Thermo Scientific™
5043B |
0.43 μm | 100 mL |
Each for $4,644.38
|
|
||||
09980295
|
Thermo Scientific™
5043C |
0.43 μm | 1000 mL |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
09980299
|
Thermo Scientific™
5049A |
0.49 μm | 15 mL |
Each for $1,189.53
|
|
||||
09980300
|
Thermo Scientific™
5049B |
0.49 μm | 100 mL |
Each for $4,644.38
|
|
||||
09980301
|
Thermo Scientific™
5049C |
0.49 μm | 1000 mL |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
09980302
|
Thermo Scientific™
5050A |
0.5 μm | 15 mL |
Each for $1,189.53
|
|
||||
09980303
|
Thermo Scientific™
5050B |
0.5 μm | 100 mL |
Each for $4,644.38
|
|
||||
09980304
|
Thermo Scientific™
5050C |
0.5 μm | 1000 mL |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
09980305
|
Thermo Scientific™
5051A |
0.51 μm | 15 mL |
Each for $1,189.53
|
|
||||
09980306
|
Thermo Scientific™
5051B |
0.51 μm | 100 mL |
Each for $4,644.38
|
|
||||
09980307
|
Thermo Scientific™
5051C |
0.51 μm | 1000 mL |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
09980308
|
Thermo Scientific™
5052A |
0.52 μm | 15 mL |
Each for $1,189.53
|
|
||||
09980309
|
Thermo Scientific™
5052B |
0.52 μm | 100 mL |
Each for $4,644.38
|
|
||||
09980311
|
Thermo Scientific™
5060A |
0.6 μm | 15 mL |
Each for $1,189.53
|
|
||||
09980312
|
Thermo Scientific™
5060B |
0.6 μm | 100 mL |
Each for $4,644.38
|
|
||||
09980313
|
Thermo Scientific™
5060C |
0.6 μm | 1000 mL |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
09980315
|
Thermo Scientific™
5065B |
0.65 μm | 100 mL |
Each for $4,644.38
|
|
||||
09980316
|
Thermo Scientific™
5065C |
0.65 μm | 1000 mL |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
09980317
|
Thermo Scientific™
5067A |
0.67 μm | 15 mL |
Each for $1,189.53
|
|
||||
09980318
|
Thermo Scientific™
5067B |
0.67 μm | 100 mL |
Each for $4,644.38
|
|
||||
09980319
|
Thermo Scientific™
5067C |
0.67 μm | 1000 mL |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
09-982-192
|
Thermo Scientific™
5074A |
0.75 μm | 15 mL |
Each for $1,189.53
|
|
||||
09-982-193
|
Thermo Scientific™
5074B |
0.75 μm | 100 mL |
Each for $4,644.38
|
|
||||
09980323
|
Thermo Scientific™
5081A |
0.82 μm | 15 mL |
Each for $1,319.19
|
|
||||
09980324
|
Thermo Scientific™
5081B |
0.82 μm | 100 mL |
Each for $4,644.38
|
|
||||
09980325
|
Thermo Scientific™
5081C |
0.82 μm | 1000 mL |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
09980326
|
Thermo Scientific™
5088A |
0.87 μm | 15 mL |
Each for $1,319.19
|
|
||||
09980327
|
Thermo Scientific™
5088B |
0.87 μm | 100 mL |
Each for $4,644.38
|
|
||||
09980328
|
Thermo Scientific™
5088C |
0.87 μm | 1000 mL |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
09980329
|
Thermo Scientific™
5093A |
0.93 μm | 15 mL |
Each for $1,189.53
|
|
||||
09980330
|
Thermo Scientific™
5093B |
0.93 μm | 100 mL |
Each for $4,644.38
|
|
||||
09980310
|
Thermo Scientific™
5052C |
0.52 μm | 1000 mL |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
09980297
|
Thermo Scientific™
5045B |
0.45 μm | 100 mL |
Each for $4,644.38
|
|
||||
09980331
|
Thermo Scientific™
5093C |
0.93 μm | 1000 mL |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Meet the need for particulate materials with a variety of particle sizes and properties with Thermo Scientific™ 5000 Series Polymer Particle Suspensions, of special interest for research in light scattering, microporous filter checking and aerosol particle generation.
Features:
- Density of 1.05g/cm3 and a refractive index of 1.59 at 589nm
- Diameters measured by optical microscopy, photon correlation spectroscopy or light scattering
- Packaged as aqueous suspensions at 10% solids by weight
- Range up to 3.1 microns in size
- For larger particles, refer to our 7000 Series Copolymer Particle Suspensions
Ordering Information:
- 15mL, 100mL, and 1000mL bottle sizes available
- 15mL (“A” Part No. bottles), 100mL (B bottles) and 1000mL (C bottles) are packaged to order; example (Part No. 5003B and 5003C).
- Please allow 10 working days when ordering these fill volumes.
Includes:
Material Safety Data Sheet (MSDS) and general product handling insert sheet
Recommended for:
High concentration polymer particles < 3μm used in filter evaluation and testing, fluid mechanics research, dispersion studies and many other research and development projects. Not intended for use in instrument calibration or diagnostic reagents because they lack the exacting specifications needed for those applications.
Specifications
Contains trace amount of surfactant to inhibit agglomeration and promote stability | |
Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer. | |
White or Colorless | |
Polymer Particle Suspensions | |
1.05g/cm3 | |
Product stable for a minimum of 24 months |
10% | |
Particles useful for such applications as filter evaluation, checking and testing, light scattering research, fluid mechanics research, aerosol particle generation, dispersion studies, and many other research and development projects. They are not intended for use in instrument calibration. | |
Polystyrene | |
General product handling insert sheet | |
1.59 at 589nm (25°C) |