Honeywell™ Miller™ AirCore™ Steel Harnesses
Comes in a high-visibility color for greater safety.
Supplier: Honeywell™ ACTBBDPUGN
Description
- Lightweight, unique breathable, open-core padding technology provides optimal airflow and reduces heat and moisture build-up up
- Contoured ergonomic padding design minimizes contact around a worker's neck, lower back and inner thighs to reduce chafing and irritation
- Patented DuraFlex stretchable webbing provides greater flexibility, increased comfort
- Cam buckles provide easy one-hand adjustment
- Holds webbing firmly in place with no readjusting required
- Stand-up back D-ring easily adjusts and attaches
- Quick-connect buckles for easy donning
- Unique and vibrant patterned webbing for easy identification
- Available with quick connect (QC) or tongue buckle, side-D rings, back-D rings
- Recommended Industries/Use: Oil and Gas
- Hazards: Confined Space, fall from height
Compliance
Meets OSHA 1926.502; ANSI™ A10.32 and ANSI Z359.1 requirements.
Specifications
Side D-rings, Lumbar Pad and Removable Belt Quick-Connect Chest; Tongue Buckle Leg | |
Quick-Connect | |
Quick-Connect | |
Side |