missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Picoline, 98%

$27.23 - $130.28

Chemical Identifiers

CAS 108-89-4
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.129
MDL Number MFCD00006440
InChI Key FKNQCJSGGFJEIZ-UHFFFAOYSA-N
Synonym 4-Methylpyridine
PubChem CID 7963
ChEBI CHEBI:32547
View More Specs

Products 3 Description

Description

Applications
4-vinylpyridine is produced from 4-Picoline, also used to produce several catalysts like 4-DMAP.

Solubility
Soluble in water, alcohol and ether.

Notes
Air sensitive & hygroscopic. Store under inert gas. Store away from oxidizing agents, water & moisture.
Specifications

Chemical Identifiers

108-89-4
93.129
FKNQCJSGGFJEIZ-UHFFFAOYSA-N
7963
C6H7N
MFCD00006440
4-Methylpyridine
CHEBI:32547

Specifications

39°C (102°F)
144°C to 145°C
2500mL
108-89-4
MFCD00006440
104586
4-Methylpyridine
CC1=CC=NC=C1
93.129
CHEBI:32547
4-Picoline
Unpleasant
3°C
10 to 11
C6H7N
UN2313
14,7402
FKNQCJSGGFJEIZ-UHFFFAOYSA-N
4-methylpyridine
7963
93.13
98%
Safety and Handling

Safety and Handling

GHS H Statement
H311-H226-H302-H332-H315-H319-H335
Toxic in contact with skin.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PICOLINES

RTECSNumber : UT5425000

TSCA : TSCA