missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Methoxy-2-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 916420-86-5 |
---|---|
Molecular Formula | C9H8BrF3O |
Molecular Weight (g/mol) | 269.06 |
MDL Number | MFCD09025406 |
InChI Key | DXAUNLGVNSSAFO-UHFFFAOYSA-N |
Synonym | 4-methoxy-2-trifluoromethyl benzyl bromide, 1-bromomethyl-4-methoxy-2-trifluoromethyl benzene, 1-bromomethyl-4-methoxy-2-trifluoromethyl-benzene, 1-bromomethyl-4-methyloxy-2-trifluoromethyl benzene |
PubChem CID | 46737574 |
IUPAC Name | 1-(bromomethyl)-4-methoxy-2-(trifluoromethyl)benzene |
SMILES | COC1=CC(=C(CBr)C=C1)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH3187103
|
Thermo Scientific Chemicals
H3187103 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
916420-86-5 | |
269.06 | |
DXAUNLGVNSSAFO-UHFFFAOYSA-N | |
46737574 | |
COC1=CC(=C(CBr)C=C1)C(F)(F)F |
C9H8BrF3O | |
MFCD09025406 | |
4-methoxy-2-trifluoromethyl benzyl bromide, 1-bromomethyl-4-methoxy-2-trifluoromethyl benzene, 1-bromomethyl-4-methoxy-2-trifluoromethyl-benzene, 1-bromomethyl-4-methyloxy-2-trifluoromethyl benzene | |
1-(bromomethyl)-4-methoxy-2-(trifluoromethyl)benzene |
Specifications
916420-86-5 | |
1.5163 | |
1g | |
4-methoxy-2-trifluoromethyl benzyl bromide, 1-bromomethyl-4-methoxy-2-trifluoromethyl benzene, 1-bromomethyl-4-methoxy-2-trifluoromethyl-benzene, 1-bromomethyl-4-methyloxy-2-trifluoromethyl benzene | |
COC1=CC(=C(CBr)C=C1)C(F)(F)F | |
269.06 | |
269.06 | |
4-Methoxy-2-(trifluoromethyl)benzyl bromide |
C9H8BrF3O | |
MFCD09025406 | |
UN3265 | |
DXAUNLGVNSSAFO-UHFFFAOYSA-N | |
1-(bromomethyl)-4-methoxy-2-(trifluoromethyl)benzene | |
46737574 | |
97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures