Learn More
4-Chlorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals
$55.74 - $565.92
Chemical Identifiers
CAS | 1073-70-7 |
---|---|
Molecular Formula | C6H8Cl2N2 |
Molecular Weight (g/mol) | 179.044 |
MDL Number | MFCD00012943 |
InChI Key | YQVZREHUWCCHHX-UHFFFAOYSA-N |
Synonym | 4-chlorophenylhydrazine hydrochloride, 4-chlorophenyl hydrazine hydrochloride, p-chlorophenylhydrazine hydrochloride, p-chlorophenylhydrazine hcl, 4-chlorophenylhydrazine hcl, 1-4-chlorophenyl hydrazine hydrochloride, 4-chloro-phenyl-hydrazine hydrochloride, chlorophenylhydrazine 4-hcl, hydrazine, 4-chlorophenyl-, monohydrochloride, 4-chlorophenylhydrazine, chloride |
PubChem CID | 71600 |
IUPAC Name | (4-chlorophenyl)hydrazine;hydrochloride |
SMILES | C1=CC(=CC=C1NN)Cl.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1451806
|
Thermo Scientific Chemicals
A1451806 |
5 g |
N/A
|
|
|||||
AAA1451814
|
Thermo Scientific Chemicals
A1451814 |
25 g |
N/A
|
|
|||||
AAA1451822
|
Thermo Scientific Chemicals
A1451822 |
100 g |
N/A
|
|
|||||
Description
4-Chlorophenylhydrazine hydrochloride is used as a pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Chlorophenylhydrazine hydrochloride is used as a pharmaceutical intermediates.
Solubility
Soluble in hot water, methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
1073-70-7 | |
179.044 | |
YQVZREHUWCCHHX-UHFFFAOYSA-N | |
71600 | |
C1=CC(=CC=C1NN)Cl.Cl |
C6H8Cl2N2 | |
MFCD00012943 | |
4-chlorophenylhydrazine hydrochloride, 4-chlorophenyl hydrazine hydrochloride, p-chlorophenylhydrazine hydrochloride, p-chlorophenylhydrazine hcl, 4-chlorophenylhydrazine hcl, 1-4-chlorophenyl hydrazine hydrochloride, 4-chloro-phenyl-hydrazine hydrochloride, chlorophenylhydrazine 4-hcl, hydrazine, 4-chlorophenyl-, monohydrochloride, 4-chlorophenylhydrazine, chloride | |
(4-chlorophenyl)hydrazine;hydrochloride |
Specifications
1073-70-7 | |
Odorless | |
MFCD00012943 | |
3596254 | |
Soluble in hot water,methanol. | |
C1=CC(=CC=C1NN)Cl.Cl | |
179.044 | |
179.05 | |
4-Chlorophenylhydrazine hydrochloride |
∼216°C (decomposition) | |
C6H8Cl2N2 | |
5 g | |
4-chlorophenylhydrazine hydrochloride, 4-chlorophenyl hydrazine hydrochloride, p-chlorophenylhydrazine hydrochloride, p-chlorophenylhydrazine hcl, 4-chlorophenylhydrazine hcl, 1-4-chlorophenyl hydrazine hydrochloride, 4-chloro-phenyl-hydrazine hydrochloride, chlorophenylhydrazine 4-hcl, hydrazine, 4-chlorophenyl-, monohydrochloride, 4-chlorophenylhydrazine, chloride | |
YQVZREHUWCCHHX-UHFFFAOYSA-N | |
(4-chlorophenyl)hydrazine;hydrochloride | |
71600 | |
97% |
Safety and Handling
GHS H Statement
H301-H350-H315-H319-H335
Toxic if swallowed.
May cause cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
EINECSNumber : 214-030-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only