Learn More
4,4'-Dimethoxybenzhydrol, 98+%, Thermo Scientific Chemicals
$139.81 - $472.18
Chemical Identifiers
CAS | 728-87-0 |
---|---|
Molecular Formula | C15H16O3 |
Molecular Weight (g/mol) | 244.29 |
MDL Number | MFCD00008410 |
InChI Key | ZODAOVNETBTTJX-UHFFFAOYSA-N |
Synonym | 4,4'-dimethoxybenzhydrol, bis 4-methoxyphenyl methanol, p,p'-dimethoxybenzhydryl alcohol, p,p'-dimethoxybenzhydrol, bis p-methoxyphenyl carbinol, benzenemethanol, 4-methoxy-.alpha.-4-methoxyphenyl, 4-methoxy-4'-methoxybenzhydrol, bis 4-methoxyphenyl methan-1-ol, benzhydrol, 4,4'-dimethoxy, acmc-209ooh |
PubChem CID | 69768 |
IUPAC Name | bis(4-methoxyphenyl)methanol |
SMILES | COC1=CC=C(C=C1)C(O)C1=CC=C(OC)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1460409
|
Thermo Scientific Chemicals
A1460409 |
10 g |
Each for $139.81
|
|
|||||
AAA1460418
|
Thermo Scientific Chemicals
A1460418 |
50 g |
Each for $472.18
|
|
|||||
Description
4,4?-Dimethoxybenzhydrol was used to study the kinetics and mechanism of oxidation of substituted benzhydrols to corresponding benzophenones in the presence of Hg(OAc)2 by N-bromosuccinimide. It is useful in the protection of glutamine and asparagine residues in peptide synthesis, reagent for the N-protection of amides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4,4′-Dimethoxybenzhydrol was used to study the kinetics and mechanism of oxidation of substituted benzhydrols to corresponding benzophenones in the presence of Hg(OAc)2 by N-bromosuccinimide. It is useful in the protection of glutamine and asparagine residues in peptide synthesis, reagent for the N-protection of amides.
Solubility
Soluble in water (0.44 g/L @ 25°C), methanol (0.1 g/mL).
Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
728-87-0 | |
244.29 | |
ZODAOVNETBTTJX-UHFFFAOYSA-N | |
69768 | |
COC1=CC=C(C=C1)C(O)C1=CC=C(OC)C=C1 |
C15H16O3 | |
MFCD00008410 | |
4,4'-dimethoxybenzhydrol, bis 4-methoxyphenyl methanol, p,p'-dimethoxybenzhydryl alcohol, p,p'-dimethoxybenzhydrol, bis p-methoxyphenyl carbinol, benzenemethanol, 4-methoxy-.alpha.-4-methoxyphenyl, 4-methoxy-4'-methoxybenzhydrol, bis 4-methoxyphenyl methan-1-ol, benzhydrol, 4,4'-dimethoxy, acmc-209ooh | |
bis(4-methoxyphenyl)methanol |
Specifications
728-87-0 | |
C15H16O3 | |
10 g | |
4,4'-dimethoxybenzhydrol, bis 4-methoxyphenyl methanol, p,p'-dimethoxybenzhydryl alcohol, p,p'-dimethoxybenzhydrol, bis p-methoxyphenyl carbinol, benzenemethanol, 4-methoxy-.alpha.-4-methoxyphenyl, 4-methoxy-4'-methoxybenzhydrol, bis 4-methoxyphenyl methan-1-ol, benzhydrol, 4,4'-dimethoxy, acmc-209ooh | |
ZODAOVNETBTTJX-UHFFFAOYSA-N | |
bis(4-methoxyphenyl)methanol | |
69768 | |
≥98% |
69°C to 71°C | |
MFCD00008410 | |
748532 | |
Soluble in water (0.44g/L @ 25°C),methanol (0.1g/mL). | |
COC1=CC=C(C=C1)C(O)C1=CC=C(OC)C=C1 | |
244.29 | |
244.29 | |
4,4'-Dimethoxybenzhydrol |
Safety and Handling
EINECSNumber : 211-975-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only