3M™ Versaflo™TR-600 Series Filter Covers
Filter cover for use with TR- 6820N filter
Supplier: 3M™ 7100021247
Description
- Helps protect TR-6820N from physical damage
- Holds TR-6600 Prefilter or TR-662 Spark Arrestor in place
- Enables PAPR to be worn through decontamination showers or rain
- Easy to use
- 3M Catalog# TR-6800FC
Applications:
Recommended Industry: Healthcare, Pharmaceutical, Heavy Industrial, Manufacturing, General Manufacturing
Certifications
NIOSH Approved
Compliance
NIOSH approved.
Specifications
TR-6800FC Filter Cover | |
TR-6800N Filter | |
TR-6800FC | |
1/Cs. |