missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3M™ Sealed Seam Hood Assemblies
Used on 3M™ Belt-Mounted Powered Air Purifying Respirator (PAPR)s and Supplied Air Respirator Systems
$369.35 - $369.35
Specifications
Certifications/Compliance | NIOSH |
---|---|
Color | White |
Material | Tychem SL |
Product Type | Hood Assembly |
For Use With (Equipment) | Belt mounted powered air purifying respirator and supplied air respirator system |
Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
19-038908
|
3M™
7000126319 |
Collar and hardhat |
Each for $369.35
|
|
|||||
Description
- Sealed seam hood assemblies
- Offer exceptional comfort and can be worn with facial hair and glasses
- 3M Catalog# H-611
These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
Specifications
NIOSH | |
Tychem SL | |
Belt mounted powered air purifying respirator and supplied air respirator system |
White | |
Hood Assembly |