missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3M™ Folded Petroleum Sorbent
High-capacity microbundle technology with scrim on one side to increase strength and help prevent linting
Supplier: 3M™ 7000002025
Description
Can be used as a boom, sheet, pad or pillow; allowing one product to be stocked for almost any chemical spill.
- Perforations every 40cm allow required length to be easily selected
- Handy dispenser allows easy on or off-site use
These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
Specifications
31.5 gal. capacity roll | |
40 L | |
Used as a boom, sheet, pad or pillow | |
1/Pk | |
50 ft. | |
5 in. |
White | |
Roll | |
Scrim on one side to increase strength and help prevent linting; Perforations every 40cm allow required length to be easily selected; Dispenser | |
50 ft. x 5 in. (15.2 m x 12.7 cm) | |
15.2 m | |
45 cm |