3M™ Empore™ C8 and C18 Extraction Disks
Reduce extraction time and solvent use for analytical preparation of aqueous environmental samples
$577.69 - $577.69
Specifications
Certifications/Compliance EPA 506, 550.1, 525.2, 1613B, 508.1, and 3535
For Use With (Application) For extracting PCBs, PNAs, Organochlorine Pesticides, Phenoxy Acid Herbicides, Phthalates, and Triazines
Product Type Extraction disk
View More Specs
Products 1 Description
Description
Reduce extraction time and solvent use for analytical preparation of aqueous environmental samples
  • Effective for extracting PCBs, PNAs, organochlorine pesticides, phenoxy acid herbicides, phthalates, and triazines
  • Provide up to 90% solvent reduction, excellent processing speed, and contamination-free extractions with accurate and reproducible results
  • Available with a variety of packings in 47 and 90mm sizes
  • C8 disk is approved for use in EPA Method 549.1
  • C18 disks are approved for use in EPA Methods 506, 550.1, 525.2, 1613B, 508.1, and 3535; 14-386-3 is also approved for use in EPA Method 608
Specifications
Specifications
EPA 506, 550.1, 525.2, 1613B, 508.1, and 3535
For extracting PCBs, PNAs, Organochlorine Pesticides, Phenoxy Acid Herbicides, Phthalates, and Triazines
Extraction disk