Learn More
3M™ 7800S Reusable Full Facepiece
Can be used with 3M Series 2000 or 7093 filters or Series 6000 cartridges
$781.01 - $781.01
Specifications
Certifications/Compliance | NIOSH 42CFR84 |
---|---|
Connection Type | Bayonet, DIN |
Material | Silicone, Polycarbonate Lens |
For Use With (Application) | Mining, oil and gas, construction, general manufacturing, transportation |
For Use With (Equipment) | 3M™ series 2000 or 7093 filter or series 6000 cartridge |
Catalog Number | Mfr. No. | Product Type | Format | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Format | Price | Quantity | ||||
17-663C
|
3M™
7000126199 |
Full Facepiece | Small |
Each for $781.01
|
|
||||
17-663B
|
3M™
7000002045 |
Full Facepiece Respirator | Medium |
Each for $781.01
|
|
||||
17-663D
|
3M™
7000126201 |
Full Facepiece Respirator | Large |
Each for $781.01
|
|
||||
Description
When combined with appropriate attachments, these facepieces are NIOSH approved for use as air-purifying respirators, continuous-flow (Type C) airline respirators, or combination air-purifying and Type C respirators.
- Soft, pliable facepiece made of easy-to-clean silicone with a double-flap seal
- Extra-large inhalation and exhalation valves
- Six adjustable straps to distribute weight
- Standard nose cup to prevent fogging
- Large faceshield with scratch-resistant polycarbonate lens
- High-quality rust-free speaking diaphragm
- 3M Catalog# 7800S-L , 3M Catalog# 7800S-M , 3M Catalog# 7800S-S
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Order Info
To convert to a Powered Air Purifying Respirator (PAPR) system, add 3M PAPR assembly and breathing tube. Order attachments separately on www.fishersafety.com.
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Specifications
NIOSH 42CFR84 | |
Silicone, Polycarbonate Lens | |
3M™ series 2000 or 7093 filter or series 6000 cartridge |
Bayonet, DIN | |
Mining, oil and gas, construction, general manufacturing, transportation |