Thermo Scientific™ 3 LP Vortex Mixer New Lab Start-Up Promo
Choose to continuously mix or pulse samples
Manufacturer:  Thermo Scientific™ 88880017FN
Catalog No. 88-880-017FN
Description
Description
Choose to continuously mix or pulse samples
Specifications
Specifications
0 to 3,000rpm
6.8 lb.
100 to 240V
CE
US
Mixers
0.5kg (1.1 lb.)
3.1kg
50/60Hz
Continuous /Touch
2 years
8.3 x 6.1 x 3.3 in. (210 x 154 x 83mm)